Assay Details
Target Gene Details
Entrez Gene ID: | 221143 |
Gene Name: | eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 |
Gene Aliases: |
ESP13, N6AMT2 |
Location: |
Chr.13:20728931-20773958 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EEF1AKMT1 | NM_001318939.1 | NP_001305868.1 | ||
NM_174928.2 | NP_777588.1 | |||
NR_134934.1 | ||||
XM_017020432.1 | XP_016875921.1 | |||
AK316604.1 | ||||
AL597222.1 | ||||
BC018091.1 | AAH18091.1 | |||
DA514874.1 | ||||
DQ823637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951 | Chr.13:20753248 - 20779879 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
