Assay Details
Target Gene Details
| Entrez Gene ID: | 196549 | 
| Gene Name: | eukaryotic translation elongation factor 1 delta pseudogene 3 | 
| Gene Aliases: | - | 
| Location: | Chr.13:31846783-31959584 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| EEF1DP3 | NR_027062.1 | |||
| DA657873.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic