Assay Details
Target Gene Details
Entrez Gene ID: | 10161 |
Gene Name: | lysophosphatidic acid receptor 6 |
Gene Aliases: |
ARWH1, HYPT8, LAH3, P2RY5, P2Y5 |
Location: |
Chr.13:48411045-48444704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPAR6 | NM_005767.5 | NP_005758.2 | ||
BC045651.1 | AAH45651.1 |
Target Gene Details
Entrez Gene ID: | 5925 |
Gene Name: | RB transcriptional corepressor 1 |
Gene Aliases: |
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110 |
Location: |
Chr.13:48303747-48481890 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RB1 | NM_000321.2 | NP_000312.2 | ||
XM_011535171.2 | XP_011533473.1 | |||
AB208788.1 | BAD92025.1 | |||
AK291258.1 | ||||
AK299179.1 | ||||
AK300284.1 | ||||
AK307730.1 | ||||
BC039060.1 | AAH39060.1 | |||
BC040540.1 | AAH40540.1 | |||
L41870.1 | AAB59465.1 | |||
M15400.1 | AAA69807.1 | |||
M28419.1 | AAA69808.1 | |||
M33647.1 | AAA69806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832602 | Chr.13:48429023 - 48593645 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
