Assay Details
Target Gene Details
Entrez Gene ID: | 54841 |
Gene Name: | basic, immunoglobulin-like variable motif containing |
Gene Aliases: |
- |
Location: |
Chr.13:102799049-102841538 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIVM | NM_001159596.1 | NP_001153068.1 | ||
| NM_017693.3 | NP_060163.2 | |||
| AF411385.1 | AAM45761.1 | |||
| AK290658.1 | ||||
| AK304391.1 | ||||
| BC112339.1 | AAI12340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048497 | Chr.13:102768928 - 102842048 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78968156,rs79164513] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map