Assay Details
Target Gene Details
Entrez Gene ID: | 283514 |
Gene Name: | siah E3 ubiquitin protein ligase family member 3 |
Gene Aliases: |
- |
Location: |
Chr.13:45780270-45851711 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAH3 | NM_198849.2 | NP_942146.2 | ||
| BC041372.2 | AAH41372.2 | |||
| DA804734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv1024 | Chr.13:45792709 - 45837281 on Build GRCh38 | Deletion |
|
| dgv332e214 | Chr.13:45762455 - 45882541 on Build GRCh38 | Loss |
|
| nsv527222 | Chr.13:45793244 - 45810696 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map