Assay Details
Target Gene Details
Entrez Gene ID: | 79132 |
Gene Name: | DEXH-box helicase 58 |
Gene Aliases: |
D11LGP2, D11lgp2e, LGP2, RLR-3 |
Location: |
Chr.17:42101404-42112733 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX58 | NM_024119.2 | NP_077024.2 | ||
| XM_017025059.1 | XP_016880548.1 | |||
| XM_017025060.1 | XP_016880549.1 | |||
| AK021416.1 | BAB13818.1 | |||
| AK097669.1 | ||||
| AK225549.1 | ||||
| AK296840.1 | ||||
| AK300900.1 | ||||
| BC014949.1 | AAH14949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map