Assay Details
Target Gene Details
Entrez Gene ID: | 9382 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
CDG2G, LDLB |
Location: |
Chr.17:73193034-73208507 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG1 | NM_018714.2 | NP_061184.1 | ||
| AB037802.1 | BAA92619.1 | |||
| AK025633.1 | ||||
| AK098224.1 | ||||
| BC021985.2 | AAH21985.1 | |||
| BC047465.1 | AAH47465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map