Assay Details
Target Gene Details
Entrez Gene ID: | 25979 |
Gene Name: | dehydrogenase/reductase 7B |
Gene Aliases: |
CGI-93, SDR32C1 |
Location: |
Chr.17:21123369-21191972 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS7B | NM_015510.4 | NP_056325.2 | ||
XM_005256595.4 | XP_005256652.2 | |||
XM_005256596.3 | XP_005256653.1 | |||
XM_011523786.2 | XP_011522088.1 | |||
XM_017024425.1 | XP_016879914.1 | |||
XM_017024426.1 | XP_016879915.1 | |||
AF151851.1 | AAD34088.1 | |||
AL117567.1 | CAB55997.1 | |||
AY358498.2 | AAQ88862.1 | |||
BC004126.2 | AAH04126.1 | |||
BC009679.2 | AAH09679.1 | |||
BI460496.1 | ||||
CR457124.1 | CAG33405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1059210 | Chr.17:21121674 - 21207107 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
