Assay Details
Target Gene Details
Entrez Gene ID: | 57521 | 
Gene Name: | regulatory associated protein of MTOR complex 1 | 
Gene Aliases: | 
	
		KOG1, Mip1 | 
Location: | 
		
			Chr.17:80544825-80966373 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RPTOR | NM_001163034.1 | NP_001156506.1 | ||
| NM_020761.2 | NP_065812.1 | |||
| AB082951.1 | BAC06490.1 | |||
| AK055912.1 | ||||
| AK295964.1 | ||||
| AK296321.1 | ||||
| AY090663.1 | AAM09075.1 | |||
| BC064515.1 | AAH64515.1 | |||
| BC136652.1 | ||||
| BC136654.1 | 
Target Copy Number Variation Details
	    DGV Version: |  
	    Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation  |  
		            Location | CNV Subtype  |  
		            Genes | 
|---|---|---|---|
| esv24254 | Chr.17:80396528 - 80656857 on Build GRCh38 | Gain+Loss | 
							
								 | 
					
| nsv523005 | Chr.17:80552727 - 80584611 on Build GRCh38 | Loss | 
							
								 | 
					
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map