Assay Details
Target Gene Details
Entrez Gene ID: | 101928021 |
Gene Name: | uncharacterized LOC101928021 |
Gene Aliases: |
- |
Location: |
Chr.17:67244776-67262080 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928021 | NR_136398.1 | 2 | 508 | |
BI462408.1 | 2 | 471 |
