Assay Details
Target Gene Details
Entrez Gene ID: | 11325 |
Gene Name: | DEAD-box helicase 42 |
Gene Aliases: |
DDX42P, RHELP, RNAHP, SF3B8, SF3b125 |
Location: |
Chr.17:63773856-63819317 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX42 | NM_007372.3 | NP_031398.2 | ||
| NM_203499.2 | NP_987095.1 | |||
| XM_006721657.1 | XP_006721720.1 | |||
| XM_017024111.1 | XP_016879600.1 | |||
| AL527653.3 | ||||
| AL545851.3 | ||||
| BC093081.1 | AAH93081.1 | |||
| DA440475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833510 | Chr.17:63767293 - 63927790 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map