Assay Details
Target Gene Details
Entrez Gene ID: | 6352 |
Gene Name: | C-C motif chemokine ligand 5 |
Gene Aliases: |
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP |
Location: |
Chr.17:35871491-35880373 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL5 | NM_001278736.1 | NP_001265665.1 | ||
| NM_002985.2 | NP_002976.2 | |||
| AF043341.1 | AAC03541.1 | |||
| AF266753.1 | AAF73070.1 | |||
| AK312212.1 | ||||
| BC008600.1 | AAH08600.1 | |||
| CR980938.1 | ||||
| DQ230537.1 | ||||
| M21121.1 | AAA36725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map