Assay Details
Target Gene Details
Entrez Gene ID: | 83394 |
Gene Name: | PITPNM family member 3 |
Gene Aliases: |
ACKR6, CORD5, NIR1, RDGBA3 |
Location: |
Chr.17:6451263-6556557 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNM3 | NM_001165966.1 | NP_001159438.1 | ||
| NM_031220.3 | NP_112497.2 | |||
| AB209130.1 | BAD92367.1 | |||
| AF334586.1 | AAK01446.1 | |||
| AL389994.1 | CAB97544.1 | |||
| BC128583.1 | ||||
| BC128584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574278 | Chr.17:5546345 - 6722066 on Build GRCh38 | Gain |
|
| nsv952115 | Chr.17:6454781 - 6512380 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map