Assay Details
Target Gene Details
Entrez Gene ID: | 79777 |
Gene Name: | acyl-CoA binding domain containing 4 |
Gene Aliases: |
HMFT0700 |
Location: |
Chr.17:45132600-45144181 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACBD4 | NM_001321352.1 | NP_001308281.1 | ||
| NM_001321353.1 | NP_001308282.1 | |||
| XM_006722085.2 | 2 | 299 | XP_006722148.1 | |
| XM_011525259.2 | 2 | 299 | XP_011523561.1 | |
| XM_011525261.2 | 2 | 299 | XP_011523563.1 | |
| XM_017025084.1 | 2 | 299 | XP_016880573.1 | |
| XM_017025085.1 | XP_016880574.1 | |||
| XM_017025086.1 | XP_016880575.1 | |||
| XM_017025087.1 | 2 | 299 | XP_016880576.1 | |
| XM_017025088.1 | 2 | 299 | XP_016880577.1 | |
| XM_017025089.1 | 2 | 299 | XP_016880578.1 | |
| XM_017025090.1 | 2 | 299 | XP_016880579.1 | |
| XM_017025091.1 | 2 | 299 | XP_016880580.1 | |
| XM_017025092.1 | XP_016880581.1 | |||
| XM_017025093.1 | 2 | 299 | XP_016880582.1 | |
| XM_017025095.1 | 2 | 299 | XP_016880584.1 | |
| XM_017025096.1 | 2 | 299 | XP_016880585.1 | |
| XM_017025097.1 | 2 | 299 | XP_016880586.1 | |
| XM_017025098.1 | XP_016880587.1 | |||
| XM_017025099.1 | 2 | 299 | XP_016880588.1 | |
| XM_017025100.1 | 2 | 299 | XP_016880589.1 | |
| XM_017025101.1 | 2 | 299 | XP_016880590.1 | |
| BC041143.1 | AAH41143.1 | |||
| DA039654.1 | 2 | 276 | ||
| DB128538.1 | 2 | 299 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2666936 | Chr.17:45134343 - 45140631 on Build GRCh38 | Deletion |
|
| nsv953903 | Chr.17:45081533 - 45148633 on Build GRCh38 | Deletion |
|
| nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map