Assay Details
Target Gene Details
Entrez Gene ID: | 1630 |
Gene Name: | DCC netrin 1 receptor |
Gene Aliases: |
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1 |
Location: |
Chr.18:52340172-53535903 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCC | NM_005215.3 | NP_005206.2 | ||
| XM_011525843.1 | XP_011524145.1 | |||
| XM_017025568.1 | XP_016881057.1 | |||
| XM_017025569.1 | XP_016881058.1 | |||
| M32292.1 | AAA35751.1 | |||
| X76132.1 | CAA53735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583063 | Chr.18:52354448 - 52356219 on Build GRCh38 | Loss |
|
| esv275550 | Chr.18:52353872 - 52355816 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72914993] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map