Assay Details
Target Gene Details
Entrez Gene ID: | 2589 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
GALNAC-T1 |
Location: |
Chr.18:35581066-35711834 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT1 | XM_017025692.1 | XP_016881181.1 | ||
| AK293062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065113 | Chr.18:35441391 - 36065348 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72958830] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map