Assay Details
Target Gene Details
Entrez Gene ID: | 9063 |
Gene Name: | protein inhibitor of activated STAT 2 |
Gene Aliases: |
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4 |
Location: |
Chr.18:46803224-46920167 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIAS2 | NM_001324046.1 | 16 | 7676 | NP_001310975.1 |
| NM_001324047.1 | 15 | 7616 | NP_001310976.1 | |
| NM_001324048.1 | 16 | 7702 | NP_001310977.1 | |
| NM_001324049.1 | 17 | 7825 | NP_001310978.1 | |
| NM_004671.4 | 14 | 7504 | NP_004662.2 | |
| NR_136684.1 | 13 | 7419 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428355 | Chr.18:46580218 - 46826322 on Build GRCh38 | Gain |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| dgv614e214 | Chr.18:46702717 - 46833888 on Build GRCh38 | Gain |
|
| nsv1056282 | Chr.18:46771152 - 46808270 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map