Assay Details
Target Gene Details
Entrez Gene ID: | 8774 |
Gene Name: | NSF attachment protein gamma |
Gene Aliases: |
GAMMASNAP |
Location: |
Chr.18:10525876-10552769 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPG | NM_003826.2 | NP_003817.1 | ||
| XM_011525754.2 | XP_011524056.1 | |||
| XM_011525756.2 | XP_011524058.1 | |||
| AK294038.1 | ||||
| AK314495.1 | ||||
| BC001889.1 | AAH01889.1 | |||
| CR536554.1 | CAG38791.1 | |||
| DA296597.1 | ||||
| U78107.1 | AAB69753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059110 | Chr.18:10142986 - 10596015 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116662191] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map