Assay Details
Target Gene Details
Entrez Gene ID: | 1000 |
Gene Name: | cadherin 2 |
Gene Aliases: |
CD325, CDHN, CDw325, NCAD |
Location: |
Chr.18:27932878-28177446 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH2 | XM_017025514.1 | XP_016881003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059472 | Chr.18:27276596 - 28749090 on Build GRCh38 | Gain |
![]() ![]() |
esv2761996 | Chr.18:27286119 - 28238654 on Build GRCh38 | Loss |
![]() ![]() |
esv2760473 | Chr.18:27806124 - 27991502 on Build GRCh38 | Gain |
![]() |
esv3573931 | Chr.18:27677585 - 28088857 on Build GRCh38 | Gain |
![]() |
nsv833605 | Chr.18:27887132 - 28063483 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
