Assay Details
Target Gene Details
Entrez Gene ID: | 9388 |
Gene Name: | lipase G, endothelial type |
Gene Aliases: |
EDL, EL, PRO719 |
Location: |
Chr.18:49560699-49592908 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPG | XM_005258390.1 | 1 | 361 | XP_005258447.1 |
| XM_011526267.1 | 1 | 361 | XP_011524569.1 | |
| XM_017026095.1 | 1 | 361 | XP_016881584.1 | |
| AK125344.1 | 1 | 361 |
More Information
Set Membership: |
|

Genomic Map