Assay Details
Target Gene Details
Entrez Gene ID: | 4089 |
Gene Name: | SMAD family member 4 |
Gene Aliases: |
DPC4, JIP, MADH4, MYHRS |
Location: |
Chr.18:51030213-51085042 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD4 | NM_005359.5 | NP_005350.1 | ||
| AK290770.1 | ||||
| BC002379.2 | AAH02379.1 | |||
| KU178168.1 | ||||
| KU178169.1 | ||||
| U44378.1 | AAA91041.1 |
More Information
Set Membership: |
|

Genomic Map