Assay Details
Target Gene Details
Entrez Gene ID: | 23327 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase |
Gene Aliases: |
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2 |
Location: |
Chr.18:58044362-58401540 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEDD4L | NM_001144964.1 | NP_001138436.1 | ||
| NM_001144967.2 | NP_001138439.1 | |||
| NM_001243960.1 | NP_001230889.1 | |||
| NM_015277.5 | NP_056092.2 | |||
| XM_006722426.3 | XP_006722489.1 | |||
| XM_006722428.3 | XP_006722491.1 | |||
| XM_017025678.1 | XP_016881167.1 | |||
| AB071179.1 | BAB69424.1 | |||
| AF385931.1 | AAM46208.1 | |||
| AY243317.1 | 1 | 72 | ||
| AY312514.1 | AAP75706.1 | |||
| BC032597.1 | AAH32597.1 | |||
| DB473368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059990 | Chr.18:58038174 - 58063549 on Build GRCh38 | Gain |
|
| dgv3360n100 | Chr.18:58025153 - 58063549 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75681926] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map