Assay Details
Target Gene Details
Entrez Gene ID: | 400794 |
Gene Name: | uncharacterized LOC400794 |
Gene Aliases: |
- |
Location: |
Chr.1:165476842-165582155 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400794 | NR_026744.2 | |||
| AK093132.1 |

Genomic Map