Assay Details
Target Gene Details
Entrez Gene ID: | 84950 |
Gene Name: | pre-mRNA processing factor 38A |
Gene Aliases: |
Prp38 |
Location: |
Chr.1:52404547-52419723 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38A | NM_032864.3 | NP_116253.2 | ||
| XM_011542315.2 | XP_011540617.1 | |||
| AK027842.1 | BAB55405.1 | |||
| AK092038.1 | BAC03796.1 | |||
| BC000777.2 | AAH00777.2 | |||
| BC063655.1 | AAH63655.1 | |||
| BC105004.1 | AAI05005.1 | |||
| BC105006.1 | AAI05007.1 |
More Information
Set Membership: |
|

Genomic Map