Assay Details
Target Gene Details
Entrez Gene ID: | 80135 |
Gene Name: | ribosome production factor 1 homolog |
Gene Aliases: |
BXDC5 |
Location: |
Chr.1:84479237-84498350 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPF1 | NM_025065.6 | NP_079341.2 | ||
AF322053.1 | AAL37236.1 | |||
AK022537.1 | BAB14086.1 | |||
BC016051.1 | AAH16051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830437 | Chr.1:84371841 - 84535586 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
