Assay Details
Target Gene Details
Entrez Gene ID: | 10207 | 
Gene Name: | PATJ, crumbs cell polarity complex component | 
Gene Aliases: | 
	
		Cipp, INADL, InaD-like, hINADL | 
Location: | 
		
			Chr.1:61742477-62163920 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PATJ | NM_176877.2 | NP_795352.2 | ||
| XM_005270341.3 | XP_005270398.1 | |||
| XM_005270347.1 | XP_005270404.1 | |||
| XM_006710278.3 | XP_006710341.1 | |||
| XM_011540462.2 | XP_011538764.1 | |||
| XM_011540464.2 | XP_011538766.1 | |||
| XM_011540465.2 | XP_011538767.1 | |||
| XM_011540466.2 | XP_011538768.1 | |||
| XM_011540467.2 | XP_011538769.1 | |||
| XM_011540468.2 | XP_011538770.1 | |||
| XM_011540469.2 | XP_011538771.1 | |||
| XM_011540470.1 | XP_011538772.1 | |||
| XM_016999998.1 | XP_016855487.1 | |||
| XM_016999999.1 | XP_016855488.1 | |||
| AJ001306.1 | CAA04666.1 | |||
| AJ224747.1 | CAA12112.1 | |||
| AJ224748.1 | CAA12113.1 | |||
| AK307713.1 | ||||
| AK308428.1 | ||||
| BC142661.1 | ||||
| BC142723.1 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map