Assay Details
Target Gene Details
| Entrez Gene ID: | 55001 | 
| Gene Name: | tetratricopeptide repeat domain 22 | 
| Gene Aliases: | - | 
| Location: | Chr.1:54779696-54802379 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| TTC22 | NM_001114108.1 | NP_001107580.1 | ||
| NM_017904.3 | NP_060374.2 | |||
| XM_011541671.2 | XP_011539973.1 | |||
| XM_017001582.1 | XP_016857071.1 | |||
| AK000626.1 | BAA91293.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| dgv166n100 | Chr.1:54741809 - 54804866 on Build GRCh38 | Gain |  MROH7-TTC4  PARS2  TTC22  TTC4 | 
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |  ACOT11  MROH7-TTC4  LEXM  PARS2  TTC22  TTC4  MROH7 | 
| nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |  ACOT11  FAM151A  MROH7-TTC4  LEXM  PARS2  SSBP3  DHCR24  TTC22  TTC4  MROH7 | 
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |  ACOT11  MROH7-TTC4  LEXM  PARS2  TTC22  TTC4  MROH7 | 
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |  FAM151A  PARS2  BSND  LOC100507634  DHCR24  USP24  ACOT11  MROH7-TTC4  LEXM  MIR4422  PCSK9  TMEM61  TTC22  TTC4  MROH7 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic