Assay Details
Target Gene Details
Entrez Gene ID: | 25932 |
Gene Name: | chloride intracellular channel 4 |
Gene Aliases: |
CLIC4L, H1, MTCLIC, huH1, p64H1 |
Location: |
Chr.1:24745269-24844324 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIC4 | NM_013943.2 | NP_039234.1 | ||
| AF097330.1 | AAD38446.1 | |||
| AF109196.1 | AAD26136.1 | |||
| AK001624.1 | BAA91794.1 | |||
| AK095959.1 | ||||
| AK301466.1 | ||||
| AK307142.1 | ||||
| AK315051.1 | ||||
| AL117424.1 | CAB55916.1 | |||
| BC012444.1 | AAH12444.1 | |||
| CR533501.1 | CAG38532.1 | |||
| DA904028.1 | ||||
| DB091421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522437 | Chr.1:24754031 - 24860225 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map