Assay Details
Target Gene Details
Entrez Gene ID: | 26092 |
Gene Name: | torsin 1A interacting protein 1 |
Gene Aliases: |
LAP1, LAP1B |
Location: |
Chr.1:179882042-179920077 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR1AIP1 | NM_001267578.1 | NP_001254507.1 | ||
| NM_015602.3 | NP_056417.2 | |||
| XM_011509403.1 | XP_011507705.1 | |||
| XM_011509404.1 | XP_011507706.1 | |||
| AK001780.1 | BAA91906.1 | |||
| AK021613.1 | BAB13855.1 | |||
| AK023204.1 | BAB14461.1 | |||
| AK291495.1 | ||||
| AK310012.1 | ||||
| BC023247.1 | AAH23247.1 | |||
| CN360425.1 |
More Information
Set Membership: |
|

Genomic Map