Assay Details
Target Gene Details
Entrez Gene ID: | 5586 |
Gene Name: | protein kinase N2 |
Gene Aliases: |
PAK2, PRK2, PRKCL2, PRO2042, Pak-2, STK7 |
Location: |
Chr.1:88684144-88836255 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKN2 | NM_001320707.1 | NP_001307636.1 | ||
| NM_001320708.1 | NP_001307637.1 | |||
| NM_001320709.1 | NP_001307638.1 | |||
| NM_006256.3 | NP_006247.1 | |||
| XM_017001782.1 | XP_016857271.1 | |||
| XM_017001783.1 | XP_016857272.1 | |||
| AF118089.2 | AAF22033.1 | |||
| AK298595.1 | ||||
| AK300304.1 | ||||
| BC125199.1 | ||||
| DA451427.1 | ||||
| S75548.1 | AAB33346.1 | |||
| U33052.1 | AAC50208.1 |
More Information
Set Membership: |
|

Genomic Map