Assay Details
Target Gene Details
Entrez Gene ID: | 8458 |
Gene Name: | transcription termination factor 2 |
Gene Aliases: |
HuF2, ZGRF6 |
Location: |
Chr.1:117060288-117107453 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTF2 | NM_003594.3 | NP_003585.3 | ||
XM_005271277.3 | XP_005271334.1 | |||
XM_011542303.2 | XP_011540605.1 | |||
XM_017002549.1 | XP_016858038.1 | |||
XM_017002550.1 | XP_016858039.1 | |||
XM_017002551.1 | XP_016858040.1 | |||
XM_017002552.1 | XP_016858041.1 | |||
XM_017002553.1 | XP_016858042.1 | |||
XM_017002554.1 | XP_016858043.1 | |||
XM_017002555.1 | XP_016858044.1 | |||
AF073771.1 | AAC64044.1 | |||
AF080255.1 | AAD49435.1 | |||
AK291017.1 | ||||
BC030058.1 | AAH30058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
