Assay Details
Target Gene Details
Entrez Gene ID: | 2752 |
Gene Name: | glutamate-ammonia ligase |
Gene Aliases: |
GLNS, GS, PIG43, PIG59 |
Location: |
Chr.1:182378093-182392206 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLUL | NM_001033044.3 | 7 | 6893 | NP_001028216.1 |
| NM_001033056.3 | 7 | 6595 | NP_001028228.1 | |
| NM_002065.6 | 8 | 7249 | NP_002056.2 | |
| BI668874.1 | 1 | 130 | ||
| CD300750.1 | 1 | 149 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114149188,rs114724499] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map