Assay Details
Target Gene Details
Entrez Gene ID: | 51742 |
Gene Name: | AT-rich interaction domain 4B |
Gene Aliases: |
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180 |
Location: |
Chr.1:235166895-235328240 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID4B | NM_001206794.1 | NP_001193723.1 | ||
| NM_016374.5 | NP_057458.4 | |||
| NM_031371.3 | NP_112739.2 | |||
| NR_135074.1 | ||||
| XM_006711781.2 | XP_006711844.1 | |||
| XM_011544212.2 | XP_011542514.1 | |||
| XM_017001468.1 | XP_016856957.1 | |||
| XM_017001469.1 | XP_016856958.1 | |||
| XM_017001470.1 | XP_016856959.1 | |||
| XM_017001471.1 | XP_016856960.1 | |||
| XM_017001472.1 | XP_016856961.1 | |||
| AB210032.1 | ||||
| AF083249.1 | AAD41239.1 | |||
| AF208045.2 | AAF23433.3 | |||
| AF214114.2 | AAF28341.2 | |||
| AF227899.1 | AAF36964.1 | |||
| AK023144.1 | BAB14428.1 | |||
| AY220790.1 | AAO63590.1 | |||
| BC130418.1 | ||||
| BX648820.1 | CAI46047.1 |
More Information
Set Membership: |
|

Genomic Map