Assay Details
Target Gene Details
Entrez Gene ID: | 1912 |
Gene Name: | polyhomeotic homolog 2 |
Gene Aliases: |
EDR2, HPH2, PH2 |
Location: |
Chr.1:33323623-33431017 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834613 | Chr.1:33334605 - 33496188 on Build GRCh38 | Gain |
|
| nsv516693 | Chr.1:33319026 - 33354533 on Build GRCh38 | Loss |
|
| nsv522364 | Chr.1:33266591 - 33367332 on Build GRCh38 | Loss |
|
| nsv545963 | Chr.1:33254041 - 33354533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map