Assay Details
Target Gene Details
Entrez Gene ID: | 79739 |
Gene Name: | tubulin tyrosine ligase like 7 |
Gene Aliases: |
- |
Location: |
Chr.1:83865021-83999340 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL7 | NM_024686.4 | NP_078962.4 | ||
| XM_005271208.2 | XP_005271265.1 | |||
| XM_017002347.1 | XP_016857836.1 | |||
| XM_017002348.1 | XP_016857837.1 | |||
| XM_017002349.1 | XP_016857838.1 | |||
| AK026686.1 | BAB15526.1 | |||
| AK094174.1 | ||||
| AK126792.1 | BAC86695.1 | |||
| AK128604.1 | BAC87524.1 | |||
| AK225574.1 | ||||
| AY170843.1 | AAO37763.1 | |||
| BC028350.2 | AAH28350.2 |
More Information
Set Membership: |
|

Genomic Map