Assay Details
Target Gene Details
Entrez Gene ID: | 284649 |
Gene Name: | SMG7 antisense RNA 1 |
Gene Aliases: |
DKFZP564C196 |
Location: |
Chr.1:183460874-183471982 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMG7-AS1 | NR_040063.1 |

Genomic Map