Assay Details
Target Gene Details
Entrez Gene ID: | 10026 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class K |
Gene Aliases: |
GPI8 |
Location: |
Chr.1:77088981-77219447 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGK | NM_005482.2 | NP_005473.1 | ||
| AF022913.1 | AAB81597.1 | |||
| AK304340.1 | ||||
| AK313015.1 | ||||
| BC020737.1 | AAH20737.1 | |||
| BX648473.1 | ||||
| Y07596.1 | CAA68871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586517 | Chr.1:77113343 - 77115270 on Build GRCh38 | Loss |
|
| esv3586515 | Chr.1:77109467 - 77113856 on Build GRCh38 | Loss |
|
| nsv830292 | Chr.1:76959886 - 77118622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map