Assay Details
Target Gene Details
Entrez Gene ID: | 9122 |
Gene Name: | solute carrier family 16 member 4 |
Gene Aliases: |
MCT4, MCT5 |
Location: |
Chr.1:110362851-110391970 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC16A4 | NM_001201546.1 | NP_001188475.1 | ||
NM_001201547.1 | NP_001188476.1 | |||
NM_001201548.1 | NP_001188477.1 | |||
NM_001201549.1 | NP_001188478.1 | |||
NM_001319220.1 | NP_001306149.1 | |||
NM_004696.2 | NP_004687.1 | |||
XM_005271317.4 | XP_005271374.1 | |||
XM_006711033.3 | XP_006711096.1 | |||
AK129985.1 | ||||
AK223307.1 | BAD97027.1 | |||
AK290720.1 | ||||
AK295946.1 | ||||
AK298539.1 | ||||
AK313735.1 | ||||
AL833619.1 | ||||
BC021664.1 | AAH21664.1 | |||
BX647948.1 | ||||
U59185.1 | AAB72035.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
