Assay Details
Target Gene Details
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLOD1 | NM_000302.3 | NP_000293.2 | ||
NM_001316320.1 | NP_001303249.1 | |||
AK294688.1 | ||||
AK295254.1 | ||||
AK299150.1 | ||||
AK312243.1 | ||||
AK316285.1 | ||||
AU139472.1 | ||||
BC016657.2 | AAH16657.1 | |||
DC334246.1 | ||||
M98252.1 |

More Information
Additional Information:
For this assay, SNP(s) [rs77367291] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
