Assay Details
Target Gene Details
| Entrez Gene ID: | 22823 | 
| Gene Name: | metal response element binding transcription factor 2 | 
| Gene Aliases: | M96, PCL2, TDRD19A, dJ976O13.2 | 
| Location: | Chr.1:93079219-93139081 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MTF2 | NM_001164391.1 | NP_001157863.1 | ||
| NM_001164392.1 | NP_001157864.1 | |||
| NM_001164393.1 | NP_001157865.1 | |||
| NM_007358.3 | NP_031384.1 | |||
| XM_011541016.1 | XP_011539318.1 | |||
| XM_011541017.2 | XP_011539319.1 | |||
| XM_011541019.1 | XP_011539321.1 | |||
| XM_017000676.1 | XP_016856165.1 | |||
| XM_017000677.1 | XP_016856166.1 | |||
| XM_017000678.1 | XP_016856167.1 | |||
| XM_017000679.1 | XP_016856168.1 | |||
| AF086908.1 | AAP97138.1 | |||
| AJ010014.1 | CAA08970.1 | |||
| AK290318.1 | ||||
| AK302776.1 | ||||
| AK302900.1 | ||||
| BC010013.2 | AAH10013.1 | |||
| DB173315.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv508404 | Chr.1:93081826 - 93114455 on Build GRCh38 | Deletion |  MTF2 | 
| nsv1008602 | Chr.1:92711254 - 93175251 on Build GRCh38 | Gain |  FAM69A  EVI5  RPL5  SNORD21  TMED5  SNORA66  MTF2 | 
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |  CCDC18  TMED5  MTF2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top