Assay Details
Target Gene Details
Entrez Gene ID: | 51611 |
Gene Name: | diphthamide biosynthesis 5 |
Gene Aliases: |
AD-018, CGI-30, HSPC143, NPD015 |
Location: |
Chr.1:100989623-101025806 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH5 | NM_001077394.1 | NP_001070862.1 | ||
| NM_001077395.1 | NP_001070863.1 | |||
| NM_015958.2 | NP_057042.2 | |||
| XM_005270938.1 | XP_005270995.1 | |||
| XM_005270939.4 | XP_005270996.1 | |||
| XM_005270941.3 | XP_005270998.1 | |||
| XM_006710683.3 | XP_006710746.1 | |||
| XM_011541569.1 | XP_011539871.1 | |||
| XM_011541570.2 | XP_011539872.1 | |||
| XM_017001460.1 | XP_016856949.1 | |||
| XM_017001461.1 | XP_016856950.1 | |||
| XM_017001462.1 | XP_016856951.1 | |||
| AF132964.1 | AAD27739.1 | |||
| AF157319.1 | AAF67485.1 | |||
| AF161492.1 | AAF29107.1 | |||
| AF248965.1 | AAG44563.1 | |||
| AK125485.1 | ||||
| AK225469.1 | ||||
| AK225707.1 | ||||
| AK225951.1 | ||||
| AK289351.1 | ||||
| AK298181.1 | ||||
| BC053857.1 | AAH53857.1 | |||
| BI907367.1 | ||||
| CK002046.1 | ||||
| DA133565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map