Assay Details
Target Gene Details
Entrez Gene ID: | 93273 |
Gene Name: | LEM domain containing 1 |
Gene Aliases: |
CT50, LEMP-1 |
Location: |
Chr.1:205381378-205449956 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LEMD1 | NM_001001552.4 | NP_001001552.3 | ||
NM_001199050.1 | NP_001185979.1 | |||
NM_001199051.1 | NP_001185980.1 | |||
NM_001199052.1 | NP_001185981.1 | |||
NR_037583.1 | ||||
XM_011510160.2 | XP_011508462.1 | |||
XM_011510161.2 | XP_011508463.1 | |||
XM_011510162.2 | XP_011508464.1 | |||
XM_011510163.2 | XP_011508465.1 | |||
XM_011510164.2 | XP_011508466.1 | |||
XM_011510165.2 | XP_011508467.1 | |||
AB084764.1 | BAD20779.1 | |||
AB084765.1 | BAD20780.1 | |||
AB085610.1 | BAD20781.1 | |||
AB096677.1 | BAD20782.1 | |||
AB096678.1 | BAD20783.1 | |||
AB096679.1 | BAD20784.1 | |||
AB096680.1 | BAD20785.1 | |||
AB096681.1 | BAD20786.1 | |||
AB096682.1 | BAD20787.1 | |||
AK026628.1 | ||||
AK302392.1 | ||||
BC036636.2 | AAH36636.1 | |||
DC399934.1 |
Target Gene Details
Entrez Gene ID: | 284576 |
Gene Name: | LEMD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:205373252-205387440 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LEMD1-AS1 | NR_038425.1 | 4 | 1252 | |
AK090497.1 | 4 | 1252 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832381 | Chr.1:205290241 - 205479588 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1014629 | Chr.1:205367415 - 205435109 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
