Assay Details
Target Gene Details
Entrez Gene ID: | 5320 |
Gene Name: | phospholipase A2 group IIA |
Gene Aliases: |
MOM1, PLA2, PLA2B, PLA2L, PLA2S, PLAS1, sPLA2 |
Location: |
Chr.1:19975431-19980439 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2A | NM_000300.3 | NP_000291.1 | ||
| NM_001161727.1 | NP_001155199.1 | |||
| NM_001161728.1 | NP_001155200.1 | |||
| NM_001161729.1 | NP_001155201.1 | |||
| AK291302.1 | ||||
| AY656695.1 | AAT73043.1 | |||
| AY656696.1 | AAT73044.1 | |||
| BC005919.1 | AAH05919.1 | |||
| BM708748.1 | ||||
| BU626291.1 | ||||
| BX641714.1 | ||||
| CB132375.1 | ||||
| CR456865.1 | CAG33146.1 | |||
| M22430.1 | AAA36550.1 |
More Information
Set Membership: |
|

Genomic Map