Assay Details
Target Gene Details
Entrez Gene ID: | 7042 |
Gene Name: | transforming growth factor beta 2 |
Gene Aliases: |
LDS4, TGF-beta2 |
Location: |
Chr.1:218345334-218444619 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB2 | NM_001135599.2 | NP_001129071.1 | ||
| NM_003238.3 | NP_003229.1 | |||
| AB209842.1 | BAD93079.1 | |||
| AK295671.1 | ||||
| AK296504.1 | ||||
| AK299839.1 | ||||
| AK313106.1 | ||||
| BC096235.4 | AAH96235.1 | |||
| BC099635.1 | AAH99635.1 | |||
| M19154.1 | AAA50404.1 | |||
| Y00083.1 | CAA68279.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs145683294] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map