Assay Details
Target Gene Details
Entrez Gene ID: | 6000 |
Gene Name: | regulator of G-protein signaling 7 |
Gene Aliases: |
- |
Location: |
Chr.1:240775193-241357374 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS7 | XM_005273218.3 | 19 | 2936 | XP_005273275.1 |
XM_006711800.3 | 18 | 2727 | XP_006711863.1 | |
XM_011544246.2 | 18 | 2386 | XP_011542548.1 | |
XM_011544247.2 | 18 | 2431 | XP_011542549.1 | |
XM_017002001.1 | 18 | 2885 | XP_016857490.1 | |
XM_017002002.1 | 20 | 2811 | XP_016857491.1 | |
XM_017002003.1 | 17 | 2831 | XP_016857492.1 | |
XM_017002004.1 | 16 | 2750 | XP_016857493.1 | |
XM_017002005.1 | 18 | 3773 | XP_016857494.1 | |
XM_017002006.1 | 19 | 2803 | XP_016857495.1 | |
XM_017002007.1 | 19 | 2734 | XP_016857496.1 | |
XM_017002008.1 | 18 | 2681 | XP_016857497.1 | |
XM_017002009.1 | 16 | 2581 | XP_016857498.1 | |
XM_017002010.1 | 17 | 2366 | XP_016857499.1 | |
XM_017002011.1 | 16 | 3898 | XP_016857500.1 | |
XM_017002012.1 | 15 | 3842 | XP_016857501.1 | |
XM_017002013.1 | 14 | 3760 | XP_016857502.1 | |
AK294354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468650 | Chr.1:240682824 - 240804127 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
