Assay Details
Target Gene Details
Entrez Gene ID: | 127733 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
UBXD3 |
Location: |
Chr.1:20183106-20196060 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN10 | XM_011540699.2 | XP_011539001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|

Genomic Map