Assay Details
Target Gene Details
Entrez Gene ID: | 101929152 |
Gene Name: | P3H2 antisense RNA 1 |
Gene Aliases: |
LEPREL1-AS1 |
Location: |
Chr.3:190120964-190144846 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H2-AS1 | NR_126419.1 | |||
| BM017065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014822 | Chr.3:190065059 - 190464556 on Build GRCh38 | Gain |
|
| esv2664192 | Chr.3:190121973 - 190123370 on Build GRCh38 | Deletion |
|
| esv3599064 | Chr.3:190120192 - 190123347 on Build GRCh38 | Loss |
|
| esv2726375 | Chr.3:190121882 - 190123376 on Build GRCh38 | Deletion |
|
| esv27411 | Chr.3:190122209 - 190123188 on Build GRCh38 | Loss |
|
| esv3893843 | Chr.3:190075998 - 191094501 on Build GRCh38 | Gain |
|
| esv3599065 | Chr.3:190121907 - 190123307 on Build GRCh38 | Loss |
|

Genomic Map