Assay Details
Target Gene Details
Entrez Gene ID: | 285242 |
Gene Name: | 5-hydroxytryptamine receptor 3E |
Gene Aliases: |
5-HT3-E, 5-HT3E, 5-HT3c1 |
Location: |
Chr.3:184097064-184106995 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3E | NM_001256613.1 | NP_001243542.1 | ||
| NM_198313.2 | NP_938055.1 | |||
| AY349352.1 | AAQ93476.1 | |||
| DQ644022.1 |
More Information
Set Membership: |
|

Genomic Map