Assay Details
Target Gene Details
Entrez Gene ID: | 727936 |
Gene Name: | glucoside xylosyltransferase 2 |
Gene Aliases: |
GLT8D4 |
Location: |
Chr.3:72848522-72975374 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GXYLT2 | XM_011534067.2 | XP_011532369.1 | ||
| XM_011534068.1 | XP_011532370.1 | |||
| XM_017007123.1 | XP_016862612.1 | |||
| XM_017007124.1 | XP_016862613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763278 | Chr.3:72595871 - 73026595 on Build GRCh38 | Gain |
|
| nsv979841 | Chr.3:72885057 - 72886057 on Build GRCh38 | Duplication |
|
| nsv1012522 | Chr.3:72880784 - 72970383 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map