Assay Details
Target Gene Details
Entrez Gene ID: | 6533 |
Gene Name: | solute carrier family 6 member 6 |
Gene Aliases: |
TAUT |
Location: |
Chr.3:14402576-14489349 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A6 | NM_001134367.3 | NP_001127839.2 | ||
NM_001134368.3 | NP_001127840.1 | |||
NM_003043.5 | NP_003034.2 | |||
NR_103507.2 | ||||
XM_011534031.2 | XP_011532333.1 | |||
AB209172.1 | BAD92409.1 | |||
AK303649.1 | ||||
AK307284.1 | ||||
AK308417.1 | ||||
AK309534.1 | ||||
BC006252.2 | AAH06252.1 | |||
BC111489.1 | AAI11490.1 | |||
BC137128.1 | ||||
BC137129.1 | ||||
BM149397.1 | ||||
DC359099.1 | ||||
U16120.1 | AAA50842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834619 | Chr.3:14354326 - 14487969 on Build GRCh38 | Gain+Loss |
![]() |
esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
